2'-Bromoacetophenone 98+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: o-Bromoacetophenone

17976-0050 17976-0250 17976-1000
TS17976-0050EA 48 USD
TS17976-0050 TS17976-0250 TS17976-1000
2'-Bromoacetophenone 98+%
2'-Bromoacetophenone
Formula: C₈H₇BrO
MW: 199.05 g/mol
Boiling Pt: 116…117 °C (12 mmHg)
Density: 1.476 g/cm³
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00000067
CAS Number: 2142-69-0
EINECS: 218-398-1

Order Now

Specification Test Results

Appearance Clear light yellow to orange Liquid
Infrared spectrum Conforms
Refractive index 1.5650 to 1.5690 (20°C, 589 nm)
GC ≥98.0 %
Residue after evaporation ≤0.01 %
Iron (Fe) ≤0.2 ppm
Lead (Pb) ≤0.02 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR