(2-Methoxyphenyl)acetic acid 99%

Supplier: Thermo Scientific Chemicals
Warning

17760-0250 17760-1000
TS17760-0250EA 71 USD
TS17760-0250 TS17760-1000
(2-Methoxyphenyl)acetic acid 99%
(2-Methoxyphenyl)acetic acid
Formula: (C₆H₄OCH₃)CH₂COOH
Melting Pt: 122…125 °C
Storage Temperature: Ambient
MDL Number: MFCD00004321
CAS Number: 93-25-4
EINECS: 202-231-4

Order Now

Specification Test Results

Appearance White to cream to light pink Crystalline powder
Infrared spectrum Conforms
Melting point 122 to 125 °C
Titration with NaOH ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR