5,6,7,8-Tetrahydro-2-naphthol 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
17747-0100
17747-0500
TS17747-0100EA
45
USD
TS17747-0100
TS17747-0500
5,6,7,8-Tetrahydro-2-naphthol 98%
5,6,7,8-Tetrahydro-2-naphthol
Formula:
C₁₀H₁₂O MW: 148.2 g/mol Melting Pt: 58…62 °C Flash Pt: >112 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001738 CAS Number: 1125-78-6 EINECS: 214-410-4 |
Specification Test Results
Appearance | Beige Crystalline mass |
Infrared spectrum | Conforms |
Melting point | 58 to 62 °C |
GC | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...