2-Naphthoic acid 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: β-Naphthoic acid

17732-0250 17732-1000
TS17732-0250EA 91 USD
TS17732-0250 TS17732-1000
2-Naphthoic acid 99%
2-Naphthoic acid
Formula: C₁₁H₈O₂
MW: 172.18 g/mol
Boiling Pt: >300 °C (760 mmHg)
Melting Pt: 183…187 °C
Flash Pt: 205 °C
Storage Temperature: Ambient
MDL Number: MFCD00004101
CAS Number: 93-09-4
EINECS: 202-217-8
Merck Index: 12,06469

Order Now

Specification Test Results

Appearance White to beige Powder
Infrared spectrum Conforms
Melting point 183 to 187 °C
HPLC ≥98.5 %
Titration with NaOH ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR