3-Bromoaniline 98%

Supplier: Thermo Scientific Chemicals
Warning

17721-0050 17721-0250 17721-1000
TS17721-0050EA 33 USD
TS17721-0050 TS17721-0250 TS17721-1000
3-Bromoaniline 98%
3-Bromoaniline
Formula: C₆H₆BrN
MW: 172.02 g/mol
Boiling Pt: 251 °C (1013 hPa)
Melting Pt: 16.8 °C
Density: 1.58 g/cm³
Flash Pt: >230 °C
Storage Temperature: Ambient
MDL Number: MFCD00007757
CAS Number: 591-19-5
EINECS: 209-704-4
UN: 2810
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Clear light yellow to orange-brown Liquid
Infrared spectrum Conforms
Refractive index 1.6240 to 1.6270 (20°C, 589 nm)
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR