1,1,4,4-Tetraphenyl-1,3-butadiene 99%

Supplier: Thermo Scientific Chemicals
Warning

17654-0010 17654-0050 17654-0100 17654-0250
TS17654-0010EA 49 USD
TS17654-0010 TS17654-0050 TS17654-0100 TS17654-0250
1,1,4,4-Tetraphenyl-1,3-butadiene 99%
1,1,4,4-Tetraphenyl-1,3-butadiene
Formula: (C₆H₅)₂CCHCHC(C₆H₅)₂
MW: 358.48 g/mol
Melting Pt: 196…200 °C
Storage Temperature: Ambient
MDL Number: MFCD00004766
CAS Number: 1450-63-1
EINECS: 215-914-7

Order Now

Specification Test Results

Appearance White to light yellow Powder or needles
Infrared spectrum Conforms
Melting point 195 to 200 °C
HPLC ≥95 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR