α,α,α-Trifluoro-p-cresol 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
4-Hydroxybenzotrifluoride, 4-(Trifluoromethyl)phenol
17644-0010
17644-0050
17644-0250
TS17644-0010EA
34
USD
TS17644-0010
TS17644-0050
TS17644-0250
α,α,α-Trifluoro-p-cresol 98%
α,α,α-Trifluoro-p-cresol
Formula:
C₇H₅F₃O MW: 162.11 g/mol Boiling Pt: 71.5…72 °C (8 mmHg) Melting Pt: 40…46 °C Flash Pt: 83 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00002363 CAS Number: 402-45-9 EINECS: 206-945-7 UN: 2928 ADR: 6.1,II |
Specification Test Results
Appearance | White to yellow-brown Crystals |
Infrared spectrum | Conforms |
Melting point | 40 to 46 °C |
GC | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...