(±)-1,2,3,4-Tetrahydro-1-naphthylamine 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Tetralin-1-amine, 1,2,3,4-Tetrahydronaphthalen-1-amine

17581-0050 17581-0250
TS17581-0050EA 45 USD
TS17581-0050 TS17581-0250
(±)-1,2,3,4-Tetrahydro-1-naphthylamine 97%
(±)-1,2,3,4-Tetrahydro-1-naphthylamine
Formula: C₁₀H₁₃N
MW: 147.22 g/mol
Boiling Pt: 246…247 °C (714 mmHg)
Density: 1.02 g/cm³
Flash Pt: 112 °C
Storage Temperature: Ambient
MDL Number: MFCD00001740
CAS Number: 2217-40-5
EINECS: 218-712-7
UN: 2735

Order Now

Specification Test Results

Appearance Clear colorless to yellow-brown Liquid
Infrared spectrum Conforms
Refractive index 1.5630 to 1.5650 (20°C, 589 nm)
GC ≥96.0 %
Microbial contamination ≤5000 CFU/g (TAMC)
pH 6.5 to 7.5 (2 % in water)
Chlorides ≤2 % (as NaCl)
Calcium (Ca) ≤0.2 %
Total nitrogen 14.0 to 16.0 %
Amino nitrogen 3.0 to 5.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR