4-Hydroxy-3-methoxybenzyl alcohol 99%

Supplier: Thermo Scientific Chemicals

17458-0010 17458-0500 17458-2500
TS17458-0010EA 401 USD
TS17458-0010 TS17458-0500 TS17458-2500
4-Hydroxy-3-methoxybenzyl alcohol 99%
4-Hydroxy-3-methoxybenzyl alcohol
Formula: C₈H₁₀O₃
Melting Pt: 112…115 °C
Storage Temperature: Ambient
MDL Number: MFCD00004659
CAS Number: 498-00-0
EINECS: 207-852-4

Order Now

Specification Test Results

Appearance Off-white to beige Crystalline powder
Infrared spectrum Conforms
Melting point 112 to 115 °C
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR