4-Isopropylaniline 99%

Supplier: Thermo Scientific Chemicals
Danger

17453-0100 17453-0500
TS17453-0100EA 28 USD
TS17453-0100 TS17453-0500
4-Isopropylaniline 99%
4-Isopropylaniline
Formula: C₉H₁₃N
Boiling Pt: 226 °C (1013 hPa)
Melting Pt: –63 °C
Density: 0.989 g/cm³
Flash Pt: 92 °C
Storage Temperature: Ambient
MDL Number: MFCD00007900
CAS Number: 99-88-7
EINECS: 202-797-2
UN: 2735
ADR: 8,II
Merck Index: 13,02647

Order Now

Specification Test Results

Appearance Clear light brown to red-brown Liquid (May darken on storage)
Infrared spectrum Conforms
Refractive index 1.5420 to 1.5440 (20°C, 589 nm)
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR