4-Isopropylphenol 98%

Supplier: Thermo Scientific Chemicals
Danger

17451-0010 17451-0050 17451-1000 17451-2500
TS17451-0010EA 195 USD
TS17451-0010 TS17451-0050 TS17451-1000 TS17451-2500
4-Isopropylphenol 98%
4-Isopropylphenol
Formula: C₉H₁₂O
MW: 136.19 g/mol
Boiling Pt: 212…212.4 °C (760 mmHg)
Melting Pt: 59…62 °C
Storage Temperature: Ambient
MDL Number: MFCD00002372
CAS Number: 99-89-8
EINECS: 202-798-8
UN: 2430
ADR: 8,III

Order Now

Specification Test Results

Appearance White to pink to light brown Powder or crystals or fused solid
Infrared spectrum Conforms
Melting point 58 to 63 °C
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR