4-Methoxysalicylic acid 99%
Supplier: Thermo Scientific Chemicals
17424-0100
17424-0500
TS17424-0100EA
81
USD
TS17424-0100
TS17424-0500
4-Methoxysalicylic acid 99%
4-Methoxysalicylic acid
Formula:
C₈H₈O₄ Boiling Pt: 327 °C (1013 hPa) Melting Pt: 157…159 °C |
MDL Number:
MFCD00002450 CAS Number: 2237-36-7 EINECS: 218-801-0 |
Specification Test Results
Appearance | Off-white to beige Powder |
Infrared spectrum | Conforms |
Melting point | 157 to 161 °C |
HPLC | ≥98.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...