2-Hydroxyethyl salicylate 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
17410-0010
17410-2500
TS17410-0010EA
231
USD
TS17410-0010
TS17410-2500
2-Hydroxyethyl salicylate 98+%
2-Hydroxyethyl salicylate
Formula:
C₉H₁₀O₄ Boiling Pt: 172 °C (12 mmHg) Melting Pt: 25 °C Density: 1.25 g/cm³ Flash Pt: 174 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002862 CAS Number: 87-28-5 |
Specification Test Results
Appearance | Colorless to almost colorless Oily liquid or fused mass |
Infrared spectrum | Conforms |
Refractive index | 1.5480 to 1.5510 (20°C, 589 nm) |
GC | ≥98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...