(4-Methoxyphenyl)acetonitrile 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 4-Methoxybenzyl cyanide, 4-Methoxyphenylacetonitrile

17376-0050 17376-0250 17376-1000
TS17376-0050EA 16 USD
TS17376-0050 TS17376-0250 TS17376-1000
(4-Methoxyphenyl)acetonitrile 97%
(4-Methoxyphenyl)acetonitrile
Formula: C₉H₉NO
MW: 147.18 g/mol
Boiling Pt: 286 °C (1013 hPa)
Melting Pt: 8 °C
Density: 1.08 g/cm³
Flash Pt: 117 °C
Storage Temperature: Ambient
MDL Number: MFCD00001919
CAS Number: 104-47-2
EINECS: 203-206-0
UN: 3276
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5290 to 1.5330 (20°C, 589 nm)
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR