o-Anisic acid 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Methoxybenzoic acid

17375-0025 17375-1000 17375-5000
TS17375-0025EA 693 USD
TS17375-0025 TS17375-1000 TS17375-5000
o-Anisic acid 99%
o-Anisic acid
Formula: C₈H₈O₃
MW: 152.15 g/mol
Melting Pt: 98…102 °C
Storage Temperature: Ambient
MDL Number: MFCD00002431
CAS Number: 579-75-9
EINECS: 209-447-8

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Melting point 98 to 102 °C
HPLC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR