4,4'-Biphenol 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: [1,1'-Biphenyl]-4,4'-diol, 4,4'-Dihydroxybiphenyl, Biphenyl-4,4'-diol

17367-0100 17367-0250 17367-1000 17367-2500
TS17367-0100EA 43 USD
TS17367-0100 TS17367-0250 TS17367-1000 TS17367-2500
4,4'-Biphenol 97%
4,4'-Biphenol
Formula: C₁₂H₁₀O₂
MW: 186.21 g/mol
Melting Pt: 278…283 °C
Storage Temperature: Ambient
MDL Number: MFCD00002348
CAS Number: 92-88-6
EINECS: 202-200-5

Order Now

Specification Test Results

Appearance Off-white to light beige Powder
Infrared spectrum Conforms
Melting point 278 to 283 °C
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR