4-Bromo-2-chlorophenol 99%

Supplier: Thermo Scientific Chemicals
Danger

17353-0050 17353-0250 17353-1000
TS17353-0050EA 33 USD
TS17353-0050 TS17353-0250 TS17353-1000
4-Bromo-2-chlorophenol 99%
4-Bromo-2-chlorophenol
Formula: C₆H₄BrClO
MW: 207.45 g/mol
Boiling Pt: 232 °C (1013 hPa)
Melting Pt: 47…51 °C
Flash Pt: >112 °C
Storage Temperature: Ambient
MDL Number: MFCD00002166
CAS Number: 3964-56-5
EINECS: 223-572-5
UN: 2811
ADR: 6.1 II

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder or needles and/or chunks or low melting mass
Infrared spectrum Conforms
Melting point 47 to 51 °C
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR