2-Bromo-p-xylene 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
2-Bromo-1,4-dimethylbenzene, 2,5-Dimethylbromobenzene
17342-0250
17342-1000
TS17342-0250EA
39
USD
TS17342-0250
TS17342-1000
2-Bromo-p-xylene 97%
2-Bromo-p-xylene
Formula:
C₈H₉Br MW: 185.06 g/mol Boiling Pt: 200 °C (1013 hPa) Melting Pt: 9…10 °C Density: 1.34 g/cm³ Flash Pt: 79 °C |
MDL Number:
MFCD00000074 CAS Number: 553-94-6 EINECS: 209-055-7 |
Specification Test Results
Appearance | Clear colorless to slightly colored Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5495 to 1.5515 (20°C, 589 nm) |
GC | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...