2-Hydroxy-m-toluic acid 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 2,3-Cresotic acid, 2-Hydroxy-3-methylbenzoic acid, 3-Methylsalicylic acid

17334-0050 17334-1000 17334-5000
TS17334-0050EA 19 USD
TS17334-0050 TS17334-1000 TS17334-5000
2-Hydroxy-m-toluic acid 99%
2-Hydroxy-m-toluic acid
Formula: C₈H₈O₃
Melting Pt: 164…167 °C
Storage Temperature: Ambient
MDL Number: MFCD00002448
CAS Number: 83-40-9
EINECS: 201-473-8
Merck Index: 13,02609

Order Now

Specification Test Results

Appearance White to off-white to light pink Powder
Infrared spectrum Conforms
Melting point 163 to 167 °C
Water ≤0.3 % (K.F.)
Titration with NaOH 98.5 to 101.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR