3-Methoxy-2-nitrobenzaldehyde 97%

Supplier: Thermo Scientific Chemicals
Warning

17267-0010 17267-0050 17267-0250
TS17267-0010EA 75 USD
TS17267-0010 TS17267-0050 TS17267-0250
3-Methoxy-2-nitrobenzaldehyde 97%
3-Methoxy-2-nitrobenzaldehyde
Formula: C₈H₇NO₄
MW: 181.15 g/mol
Melting Pt: 97…101 °C
Storage Temperature: Ambient
MDL Number: MFCD00007135
CAS Number: 53055-05-3

Order Now

Specification Test Results

Appearance White to yellow to beige Crystalline powder
Infrared spectrum Conforms
Melting point 97 to 103 °C
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR