7-Methoxy-1-tetralone 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
7-Methoxy-3,4-dihydronaphthalen-1(2H)-one
17243-0050
17243-0250
TS17243-0010EA
21.34
USD
TS17243-0010
TS17243-0050
TS17243-0250
7-Methoxy-1-tetralone 97%
7-Methoxy-1-tetralone
Formula:
C₁₁H₁₂O₂ MW: 176.22 g/mol Melting Pt: 61…63 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001696 CAS Number: 6836-19-7 |
Specification Test Results
Appearance | White to beige Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 60 to 64 °C |
GC | ≥96.0 % |
Elemental Analysis | 10.41 to 11.28 % (N) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...