3-Nitrophenol 99%

Supplier: Thermo Scientific Chemicals
Danger

17230-0100 17230-0500 17230-2500
TS17230-0100EA 88 USD
TS17230-0100 TS17230-0500 TS17230-2500
3-Nitrophenol 99%
3-Nitrophenol
Formula: C₆H₅NO₃
Boiling Pt: 194 °C (70 mmHg)
Melting Pt: 94…98 °C
Density: 1.49 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00007240
CAS Number: 554-84-7
EINECS: 209-079-8
UN: 1663
ADR: 6.1,III
Merck Index: 12,06682

Order Now

Specification Test Results

Appearance Yellow to brown Crystalline powder
Infrared spectrum Conforms
Melting point 94 to 98 °C
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR