1,2-Naphthoquinone 95% tech.

Supplier: Thermo Scientific Chemicals
Warning

17123-0010 17123-0050 17123-0100 17123-0250
TS17123-0010EA 85 USD
TS17123-0010 TS17123-0050 TS17123-0100 TS17123-0250
1,2-Naphthoquinone 95% tech.
1,2-Naphthoquinone
Formula: C₁₀H₆O₂
Boiling Pt: 296 °C (1013 hPa)
Melting Pt: 136…141 °C
Storage Temperature: Ambient
MDL Number: MFCD00001698
CAS Number: 524-42-5
EINECS: 208-360-2
Merck Index: 12,06481

Order Now

Specification Test Results

Appearance Brown Powder
Infrared spectrum Conforms
GC ≥94 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR