9-Fluorenylmethanol 99%

Supplier: Thermo Scientific Chemicals
Warning

17093-0010 17093-0050 17093-0100 17093-0500
TS17093-0010EA 24 USD
TS17093-0010 TS17093-0050 TS17093-0100 TS17093-0500
9-Fluorenylmethanol 99%
9-Fluorenylmethanol
Formula: C₁₄H₁₂O
Melting Pt: 104…107 °C
Storage Temperature: Ambient
MDL Number: MFCD00001139
CAS Number: 24324-17-2
EINECS: 246-167-5

Order Now

Specification Test Results

Appearance White to light yellow Crystalline powder
Infrared spectrum Conforms
Melting point 104 to 107 °C
GC ≥98.5 %
Specific optical rotation +293° to +303° (20 °C, 589 nm) (c=2, H2O)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR