4,4'-Bis(diethylamino)benzophenone 99+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: N,N,N',N'-Tetraethyl-4,4'-diaminobenzophenone, 4,4-Bis(diethylamino) benzophenone

17081-0250 17081-1000
TS17081-0250EA 55 USD
TS17081-0250 TS17081-1000
4,4'-Bis(diethylamino)benzophenone 99+%
4,4'-Bis(diethylamino)benzophenone
Formula: C₂₁H₂₈N₂O
Boiling Pt: 420 °C (1013 hPa)
Melting Pt: 93…95 °C
Flash Pt: 151 °C
Storage Temperature: Ambient
MDL Number: MFCD00009044
CAS Number: 90-93-7
EINECS: 202-025-4
UN: 3077
ADR: 9,III

Order Now

Specification Test Results

Appearance White to yellow to yellow-green Powder or granules and/or lumps
Infrared spectrum Conforms
Melting point 93 to 96 °C
HPLC ≥99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR