3-Methoxybenzylamine 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: m-Anisylamine, 3-(Aminomethyl)anisole

17055-0100 17055-0250
TS17055-0100EA 80 USD
TS17055-0100 TS17055-0250
3-Methoxybenzylamine 98%
3-Methoxybenzylamine
Formula: C₈H₁₁NO
MW: 137.18 g/mol
Boiling Pt: 140 °C (37 mmHg)
Density: 1.07 g/cm³
Flash Pt: >110 °C
MDL Number: MFCD00008115
CAS Number: 5071-96-5
EINECS: 225-779-6
UN: 2735
ADR: 8,III

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5470 to 1.5490 (20°C, 589 nm)
GC ≥97.5 %
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR