2,4-Dimethoxybenzyl alcohol 99%

Supplier: Thermo Scientific Chemicals

17046-0250 17046-1000
TS17046-0250EA 114 USD
TS17046-0250 TS17046-1000
2,4-Dimethoxybenzyl alcohol 99%
2,4-Dimethoxybenzyl alcohol
Formula: C₉H₁₂O₃
Boiling Pt: 177…179 °C (10 mmHg)
Melting Pt: 37…40 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00004614
CAS Number: 7314-44-5
EINECS: 230-775-2

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid or low melting solid
Infrared spectrum Conforms
Melting point 34 to 38 °C
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR