Methyl-3,5-dihydroxybenzoate 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Methyl 3,5-dihydroxybenzoate, 3,5-Dihydroxybenzoic acid methyl ester

17044-0250 17044-1000
TS17044-0250EA 72 USD
TS17044-0250 TS17044-1000
Methyl-3,5-dihydroxybenzoate 97%
Methyl-3,5-dihydroxybenzoate
Formula: C₈H₈O₄
MW: 168.15 g/mol
Boiling Pt: 300 °C (1013 hPa)
Melting Pt: 164…169 °C
MDL Number: MFCD00002289
CAS Number: 2150-44-9
EINECS: 218-426-2

Order Now

Specification Test Results

Appearance Light yellow to beige Crystalline powder
Infrared spectrum Conforms
Melting point 164 to 169 °C
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR