α-Bromo-p-toluic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

17040-0100 17040-0500 17040-2500
TS17040-0100EA 96 USD
TS17040-0100 TS17040-0500 TS17040-2500
α-Bromo-p-toluic acid 97%
α-Bromo-p-toluic acid
Formula: C₈H₇BrO₂
Boiling Pt: 316 °C (1013 hPa)
Melting Pt: 223…227 °C
Storage Temperature: Ambient
MDL Number: MFCD00002567
CAS Number: 6232-88-8
EINECS: 228-343-3

Order Now

Specification Test Results

Appearance White to beige Crystalline powder
Infrared spectrum Conforms
HPLC ≥97.5 %
Titration with NaOH ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR