(Acetylmethylene)triphenylphosphorane 99%

Supplier: Thermo Scientific Chemicals

16997-0250 16997-1000
TS16997-0250EA 113 USD
TS16997-0250 TS16997-1000
(Acetylmethylene)triphenylphosphorane 99%
(Acetylmethylene)triphenylphosphorane
Formula: C₂₁H₁₉OP
Melting Pt: 203…207 °C
Storage Temperature: Ambient
MDL Number: MFCD00008774
CAS Number: 1439-36-7
EINECS: 215-878-2

Order Now

Specification Test Results

Appearance White to off-white Powder
Infrared spectrum Conforms
Melting point 203 to 211 °C
HPLC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR