Phenoxyacetic acid 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
16987-0025
16987-0050
16987-1000
16987-5000
TS16987-0025EA
245
USD
TS16987-0025
TS16987-0050
TS16987-1000
TS16987-5000
Phenoxyacetic acid 98+%
Phenoxyacetic acid
Formula:
C₈H₈O₃ Boiling Pt: 285 °C (760 mmHg) Melting Pt: 98…101 °C Flash Pt: 165 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004296 CAS Number: 122-59-8 EINECS: 204-556-7 Merck Index: 12,07407 |
Specification Test Results
Appearance | White to beige Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 98 to 102 °C |
Titration with NaOH | ≥98.0 % |
NMR (1H-NMR) | Consistent with structure |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...