4-Benzyloxyphenol 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Monobenzone4-Hydroxyphenyl benzyl etherHydroquinone monobenzyl ether4-(Benzyloxy)phenol
16978-0010
16978-0500
16978-2500
TS16978-0010EA
686
USD
TS16978-0010
TS16978-0500
TS16978-2500
4-Benzyloxyphenol 98%
4-Benzyloxyphenol
Formula:
C₁₃H₁₂O₂ MW: 200.24 g/mol Melting Pt: 121…122 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002333 CAS Number: 103-16-2 EINECS: 203-083-3 Merck Index: 12,06331 |
Specification Test Results
Appearance | Off-white to beige to brown Crystalline powder or crystals and/or chunks |
Infrared spectrum | Conforms |
Melting point | 117 to 122 °C |
GC | ≥97.5 % |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...