β-Bromostyrene (mixture of cis and trans isomers) cis- and trans-mixture 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (2-Bromovinyl)benzene, β-Bromostyrene

16929-0050 16929-0250 16929-1000
TS16929-0050EA 36 USD
TS16929-0050 TS16929-0250 TS16929-1000
β-Bromostyrene (mixture of cis and trans isomers) cis- and trans-mixture 97%
β-Bromostyrene (mixture of cis and trans isomers)
Formula: C₈H₇Br
Boiling Pt: 110…112 °C (20 mmHg)
Melting Pt: 7 °C
Density: 1.42 g/cm³
Flash Pt: 79 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000185
CAS Number: 103-64-0
EINECS: 203-131-3

Order Now

Specification Test Results

Appearance Clear yellow to brown Liquid
Infrared spectrum Conforms
Refractive index 1.6050 to 1.6090 (20°C, 589 nm)
GC ≥96.0 % (sum of isomers)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR