3-Cyanobenzoic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

16913-0100 16913-0500
TS16913-0100EA 53 USD
TS16913-0100 TS16913-0500
3-Cyanobenzoic acid 98%
3-Cyanobenzoic acid
Formula: C₈H₅NO₂
MW: 147.13 g/mol
Melting Pt: 217…222 °C
Storage Temperature: Ambient
MDL Number: MFCD00002486
CAS Number: 1877-72-1
EINECS: 217-511-1

Order Now

Specification Test Results

Appearance White to off-white Powder
Infrared spectrum Conforms
Melting point 217 to 222 °C
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR