9,10-Phenanthrenequinone 95%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Phenanthrene-9,10-dione

16879-0250 16879-1000
TS16879-0250EA 84 USD
TS16879-0250 TS16879-1000
9,10-Phenanthrenequinone 95%
9,10-Phenanthrenequinone
Formula: C₁₄H₈O₂
MW: 208.22 g/mol
Boiling Pt: 360 °C (1013 hPa)
Melting Pt: 201…211 °C
Flash Pt: 245 °C
Storage Temperature: Ambient
MDL Number: MFCD00001163
CAS Number: 84-11-7
EINECS: 201-515-5
Merck Index: 12,07355

Order Now

Specification Test Results

Appearance Orange-brown Powder
Infrared spectrum Conforms
Melting point 203 to 211 °C
HPLC ≥94 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR