3-Hydroxy-4-nitrobenzaldehyde 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
16856-0010
16856-0100
TS16856-0010EA
49
USD
TS16856-0010
TS16856-0100
3-Hydroxy-4-nitrobenzaldehyde 97%
3-Hydroxy-4-nitrobenzaldehyde
Formula:
C₇H₅NO₄ Melting Pt: 131…134 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007109 CAS Number: 704-13-2 |
Specification Test Results
Appearance | Yellow to brown Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 130 to 135 °C |
GC | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...