α,α,α-Trifluoro-m-cresol 98+%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 3-(Trifluoromethyl)phenol, 3-Hydroxybenzotrifluoride

16847-0250 16847-1000
TS16847-0250EA 27 USD
TS16847-0250 TS16847-1000
α,α,α-Trifluoro-m-cresol 98+%
α,α,α-Trifluoro-m-cresol
Formula: C₇H₅F₃O
MW: 162.11 g/mol
Boiling Pt: 178 °C (1013 hPa)
Melting Pt: –1.8 °C
Density: 1.33 g/cm³
Flash Pt: 73 °C
Storage Temperature: Ambient
MDL Number: MFCD00002299
CAS Number: 98-17-9
EINECS: 202-645-5
UN: 1760
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4570 to 1.4600 (20°C, 589 nm)
GC ≥98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR