4-Fluorostyrene 97% stabilized

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-Fluoro-4-vinylbenzene

16834-0010 16834-0100
TS16834-0010EA 31 USD
TS16834-0010 TS16834-0100
4-Fluorostyrene 97% stabilized
4-Fluorostyrene
Formula: C₈H₇F
MW: 122.14 g/mol
Boiling Pt: 145…148 °C (760 mmHg)
Melting Pt: –34.5 °C
Density: 1.024 g/cm³
Flash Pt: 26 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000361
CAS Number: 405-99-2
EINECS: 206-975-0
UN: 1993
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5140 to 1.5160 (20°C, 589 nm)
GC ≥96.0 %
Stabilizer 0.1 % tert.-butylcatechol

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR