(E)-4-Phenylbut-3-enoic acid 96%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: trans-Styrylacetic acid, trans-4-Phenylbut-3-enoic acid, trans-4-Phenyl-3-butenoic acid

16810-0010 16810-0100
TS16810-0010EA 39 USD
TS16810-0010 TS16810-0100
(E)-4-Phenylbut-3-enoic acid 96%
(E)-4-Phenylbut-3-enoic acid
Formula: C₁₀H₁₀O₂
MW: 162.188 g/mol
Melting Pt: 82…88 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00002783
CAS Number: 1914-58-5

Order Now

Specification Test Results

Appearance White to yellow-brown Crystalline powder or needles
Infrared spectrum Conforms
Melting point 82 to 88 °C
HPLC ≥95 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR