5-Bromo-o-anisaldehyde 99%

Supplier: Thermo Scientific Chemicals
Warning

16752-0050 16752-0250 16752-1000
TS16752-0050EA 24 USD
TS16752-0050 TS16752-0250 TS16752-1000
5-Bromo-o-anisaldehyde 99%
5-Bromo-o-anisaldehyde
Formula: C₈H₇BrO₂
Melting Pt: 115…118 °C
Storage Temperature: Ambient
MDL Number: MFCD00003347
CAS Number: 25016-01-7
EINECS: 246-564-3

Order Now

Specification Test Results

Appearance Off-white to yellow to beige Powder or crystals
Infrared spectrum Conforms
Melting point 115 to 119 °C
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR