4-Bromo-2-chloroaniline 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
16748-0100
16748-0500
TS16748-0100EA
44
USD
TS16748-0100
TS16748-0500
4-Bromo-2-chloroaniline 99%
4-Bromo-2-chloroaniline
Formula:
C₆H₅BrClN Boiling Pt: 264 °C (1013 hPa) Melting Pt: 67…70 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007660 CAS Number: 38762-41-3 EINECS: 254-118-4 UN: 2811 ADR: 6.1,III |
Specification Test Results
Appearance | Light yellow to light brown Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 67 to 70 °C |
GC | ≥98.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...