3-Fluorophenylhydrazinium chloride 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: (3-Fluorphenyl)hydrazin HCl, (3-Fluorophenyl)hydrazine hydrochloride, 3-Fluorophenylhydrazine hydrochloride, 1-(3-Fluorophenyl)hydrazine hydrochloride

16737-0050 16737-0100
TS16737-0050EA 74 USD
TS16737-0050 TS16737-0100
3-Fluorophenylhydrazinium chloride 97%
3-Fluorophenylhydrazinium chloride
Formula: C₆H₈ClFN₂
Melting Pt: 268 °C (dec.)
MDL Number: MFCD00012934
CAS Number: 2924-16-5
EINECS: 220-886-4
UN: 2811

Order Now

Specification Test Results

Appearance White to yellow to light brown Powder
Infrared spectrum Conforms
HPLC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR