3-Bromophenylhydrazinium chloride 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 3-Bromophenylhydrazine hydrochloride, 1-(3-Bromophenyl)hydrazine hydrochloride

16735-0050 16735-0250
TS16735-0050EA 94 USD
TS16735-0050 TS16735-0250
3-Bromophenylhydrazinium chloride 98%
3-Bromophenylhydrazinium chloride
Formula: C₆H₈BrClN₂
MW: 223.5
Melting Pt: 225…231 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00012933
CAS Number: 27246-81-7
EINECS: 248-362-0
UN: 1759
ADR: 8 II

Order Now

Specification Test Results

Appearance Off-white to light brown Powder
Infrared spectrum Conforms
Melting point 225 to 231 °C (Dec)
Titration Argentometric ≥97.5 %
Titration with NaOH 97.5 to 102.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR