4-tert-Amylphenol 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: p-(1,1-Dimethylpropyl)phenol, 4-(2-Methyl-2-butyl)phenol, 4-tert-Pentylphenol

16733-0050 16733-1000 16733-2500 16733-5000
TS16733-0050EA 24 USD
TS16733-0050 TS16733-1000 TS16733-2500 TS16733-5000
4-tert-Amylphenol 99%
4-tert-Amylphenol
Formula: C₁₁H₁₆O
MW: 164.25 g/mol
Melting Pt: >91 °C
Flash Pt: 111 °C
MDL Number: MFCD00002369
CAS Number: 80-46-6
EINECS: 201-280-9
UN: 2430
ADR: 8,III

Order Now

Specification Test Results

Appearance White to light yellow Coarse powder or flakes or briquettes
Infrared spectrum Conforms
Melting point ≥91 °C
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR