(4-Chlorophenoxy)acetic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-CPA

16696-1000 16696-5000
TS16696-1000EA 33 USD
TS16696-1000 TS16696-5000
(4-Chlorophenoxy)acetic acid 98%
(4-Chlorophenoxy)acetic acid
Formula: (C₆H₄Cl)OCH₂COOH
MW: 186.59 g/mol
Boiling Pt: 308 °C (1013 hPa)
Melting Pt: >156 °C
MDL Number: MFCD00004305
CAS Number: 122-88-3
EINECS: 204-581-3

Order Now

Specification Test Results

Appearance White to light beige Powder
Infrared spectrum Conforms
Melting point ≥156 °C
Water ≤1 %
Titration with NaOH ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR