1,4-Naphthoquinone, (max. 6% H₂O) 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
1,4-Naphthalenedione, 1,4-Dihydro-1,4-diketonaphthalene
16671-0025
16671-1000
16671-5000
TS16671-0025EA
326
USD
TS16671-0025
TS16671-1000
TS16671-5000
1,4-Naphthoquinone, (max. 6% H₂O) 99%
1,4-Naphthoquinone
Formula:
C₁₀H₆O₂ MW: 158.16 g/mol Boiling Pt: 298 Melting Pt: 124 Flash Pt: 141 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001676 CAS Number: 130-15-4 EINECS: 204-977-6 UN: 2928 ADR: 6.1 I Merck Index: 12,06482 |
Specification Test Results
Appearance | Khaki Powder |
Infrared spectrum | Conforms |
Water | ≤6 % (K.F.) |
GC | ≥99 % |
HPLC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...