4-Bromo-α,α,α-trifluorotoluene 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
4-Bromobenzotrifluoride, 1-Bromo-4-(trifluoromethyl)benzene
16667-0050
16667-0250
TS16667-0050EA
65
USD
TS16667-0050
TS16667-0250
4-Bromo-α,α,α-trifluorotoluene 99%
4-Bromo-α,α,α-trifluorotoluene
Formula:
C₇H₄BrF₃ MW: 225.01 g/mol Boiling Pt: 154…155 °C (760 mmHg) Density: 1.6 g/cm³ Flash Pt: 48 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000398 CAS Number: 402-43-7 EINECS: 206-943-6 UN: 1993 ADR: 3,III |
Specification Test Results
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4715 to 1.4745 (20°C, 589 nm) |
GC | ≥98.5 % |
HPLC | ≥98.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...