2,3,4-Trimethoxybenzaldehyde 99%

Supplier: Thermo Scientific Chemicals

16645-0100 16645-0500
TS16645-0100EA 29 USD
TS16645-0100 TS16645-0500
2,3,4-Trimethoxybenzaldehyde 99%
2,3,4-Trimethoxybenzaldehyde
Formula: C₁₀H₁₂O₄
MW: 196.2 g/mol
Boiling Pt: 168…170 °C (12 mmHg)
Melting Pt: 38…41 °C
Flash Pt: >112 °C
Storage Temperature: Ambient
MDL Number: MFCD00003310
CAS Number: 2103-57-3
EINECS: 218-271-0

Order Now

Specification Test Results

Appearance White to light yellow Crystalline powder or crystals
Infrared spectrum Conforms
Melting point ≥36.5 °C
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR