4-Methylanisole 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: p-Methylanisole

16466-0010 16466-0050 16466-2500 164660025
TS16466-0010EA 165 USD
TS16466-0010 TS16466-0050 TS16466-2500 TS164660025
4-Methylanisole 99%
4-Methylanisole
Formula: C₈H₁₀O
MW: 122.17 g/mol
Boiling Pt: 174 °C (760 mmHg)
Melting Pt: –50 °C
Density: 0.96 g/cm³
Flash Pt: 53 °C
Storage Temperature: Ambient
MDL Number: MFCD00008413
CAS Number: 104-93-8
EINECS: 203-253-7
UN: 1993
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5100 to 1.5130 (20°C, 589 nm)
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR