4,4'-Bis(dimethylamino)benzophenone 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 4,4′-Bis(dimethylamino)benzophenone, 4,4-Bis(dimethylamino)benzophenone, 4,4'-Di(dimethylamino)benzophenone, Michler's ketone, Bis[4-(dimethylamino)phenyl]methanone

16449-0050 16449-1000 16449-5000
TS16449-0050EA 22 USD
TS16449-0050 TS16449-1000 TS16449-5000
4,4'-Bis(dimethylamino)benzophenone 98%
4,4'-Bis(dimethylamino)benzophenone
Formula: C₁₇H₂₀N₂O
MW: 268.36 g/mol
Boiling Pt: 428 °C (1013 hPa)
Melting Pt: 171…176 °C
Flash Pt: 220 °C
Storage Temperature: Ambient
MDL Number: MFCD00008312
CAS Number: 90-94-8
EINECS: 202-027-5
Merck Index: 12,06265

Order Now

Specification Test Results

Appearance Blue to dark blue to grey Powder
Infrared spectrum Conforms
Melting point 171 to 176 °C (Dec)
HPLC ≥97.5 %
Titration with HClO4 ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR